Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophil Elastase Mouse anti-Human, BV421, Clone: NP57, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569513
Description
Neutrophil Elastase Monoclonal antibody specifically detects Neutrophil Elastase in Human samples. It is validated for Intracellular staining (flow cytometry)
Specifications
Neutrophil Elastase | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
1991 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 |
Flow Cytometry, Control | |
NP57 | |
Brilliant Violet 421 | |
ELANE; HNE; PMN elastase; PMN-E; neutrophil elastase | |
Neutrophil granule proteins | |
0.1 mg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction