Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophils Antibody (Nimp-R14), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280078H
This item is not returnable.
View return policy
Description
Neutrophils Monoclonal antibody specifically detects Neutrophils in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
Neutrophils | |
Monoclonal | |
HRP | |
Rat | |
Protein G purified | |
RUO | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Nimp-R14 | |
PBS | |
Purified mouse BALB/c neutrophils. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction