Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophils Antibody (Nimp-R14), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280078MFV610
Description
Neutrophils Monoclonal antibody specifically detects Neutrophils in Mouse samples. It is validated for Flow CytometrySpecifications
Neutrophils | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Purified mouse BALB/c neutrophils. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
Nimp-R14 | |
Flow Cytometry | |
Rat | |
Protein G purified | |
RUO | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction