Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophils Antibody (RM0028-3G23), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212439AF488
Description
Neutrophils Monoclonal specifically detects Neutrophils in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Neutrophils | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Isolated mouse neutrophil cells | |
100 μL | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
RM0028-3G23 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Rat | |
Protein G purified | |
Primary | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction