Learn More
Neutrophils Antibody (RM0028-3G23) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Neutrophils |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | RM0028-3G23 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1000, Immunohistochemistry 1:100-300, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:100-300 |
| Formulation | A 0.2 um filtered solution in PBS with No Preservative |
| Host Species | Rat |
| Immunogen | Isolated mouse neutrophil cells |
| Purification Method | Protein G purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.