Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutrophils Rabbit anti-Human, Mouse, Rat, Clone: FUT4/1478R, Novus Biologicals™
SDP

Catalog No. p-7079039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253367200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB253367200 Supplier Novus Biologicals Supplier No. NBP253367200UG

Rabbit Monoclonal Antibody

SSEA-1 Monoclonal antibody specifically detects SSEA-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Neutrophils
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FUT4/1478R
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 2 - 4ug/million cells, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Host Species Rabbit
Immunogen Purified human neutrophils
Molecular Weight of Antigen 220 kDa
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4C.
Form Purified
Isotype IgG κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.