Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophils Antibody (Nimp-R14) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280063
Description
Neutrophils Monoclonal specifically detects Neutrophils in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.Specifications
Neutrophils | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
Purified mouse BALB/c neutrophils. | |
500 μg | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization | |
Nimp-R14 | |
Flow Cytometry 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200, Neutralization | |
Rat | |
Protein G purified | |
RUO | |
Recognizes mouse neutrophils (surface protein mouse Ly-6G) and may recognize Ly-6Chigh cells (inflammatory monocytes). | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction