Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nevirapine, 98%

Catalog No. AC461150010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461150010 1 g
AC461152500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461150010 Supplier Thermo Scientific Chemicals Supplier No. 461150010
Add to Cart

CAS: 129618-40-2 | C15H14N4O | 266.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129618-40-2
Molecular Formula C15H14N4O
Molecular Weight (g/mol) 266.3
InChI Key NQDJXKOVJZTUJA-UHFFFAOYSA-N
Synonym nevirapine, viramune, bi-rg-587, viramune xr, nvp, birg587, 11-cyclopropyl-4-methyl-5,11-dihydro-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one, nevirapine usan:inn, unii-99dk7fvk1h, 11-cyclopropyl-5,11-dihydro-4-methyl-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one
PubChem CID 4463
ChEBI CHEBI:63613
IUPAC Name 11-cyclopropyl-4-methyl-5H-dipyrido[2,3-e
SMILES CC1=C2C(=NC=C1)N(C3=C(C=CC=N3)C(=O)N2)C4CC4

Specifications

Melting Point 247.0°C to 249.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 266.3
Percent Purity 98%
Chemical Name or Material Nevirapine
Hazard Category Acute toxicity (category 4), Hazardous to the aquatic environment (chronic category 3)
Hazard Statement Harmful if swallowed, Harmful to aquatic life with long lasting effects

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.