Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ New Brunswick Innova™ 42/42 R - Stackable Incubator Shaker PROMO

Versatile and reliable. Stackable, can be placed on or under the bench, and come with a static shelf to be used to incubate agar plates while shaking cultures in flasks (Optional: static culture drawer, gassing manifold, remote alarm, and humidity monitor and drip pan.

Specifications

Load Bearing Capacity (Metric) 9 to 15.8 kg
Speed Range 25 to 400 rpm
Electrical Requirements 120 V 50/60 Hz
Voltage 120 V
Temperature Range (Metric) 20°C Below Ambient to 80°C
View More Specs

Products 2
Catalog Number Mfr. No. Orbit Price Quantity & Availability  
Catalog Number Mfr. No. Orbit Price Quantity & Availability  
05-400-157PM
View Documents
Eppendorf™
2231001123
19 mm N/A N/A N/A
05-400-162PM
View Documents
Eppendorf™
2231001124
25 mm N/A N/A N/A
Description

Description

  • High capacity in a small footprint: for example, 4 x 5 L or 2 x 6 L flasks on universal platform; device footprint 4731 cm2 /733 in.2 (63.5 cm/25 in. width, 74.5 cm/29.3 in. depth)
  • Ergonomical and safe interface handling: Top mounted controls and display make bending down unnecessary during handling, avoids accidental parameter changes, and can be reached easily even in a double stack
  • Easy to clean: Spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation); airflow UV-germicidal light optional
  • Lab position flexibility: Lab position on or under bench, 2x stackable
  • Parallel static cultivation: One height adjustable static shelf included, optional culture drawer and additional shelves available
  • Sample evaporation protection: Bottom spill pan usable as water reservoir; optional culture drawer, for Agar plates, for example
  • Visual sample control: Large viewing window and internal light provide easy viewing of samples without the need to open the door
  • Reliable 24/7 with high loads: Proven and tested Innova triple eccentric counterbalanced drive (three weight-supporting shafts), max. load 15 kg
  • Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
  • Options and Accessories: Orbit 19 or 25 mm, photosynthetic light bank, UV germicidal light, culture drawer for evaporation protection of Agar plates, additional static shelves (one included), humidity monitoring, remote monitoring with VisioNize™ Lab Suite and more
  • Speed: 25 to 400 rpm, 25 to 300 rpm when using flasks >500 mL in stacked devices
  • Certifications/Compliance: 2014/35/EU: EN 61010-1, EN 61010-2-010, EN 61010-2-051, 2014/30/EU: EN 61326-1, EN 55011 (Class A), 2011/65/EU: EN IEC 63000, IEC 61010-1, IEC 61010-2-010, IEC 61010-2-051, UL 61010-1, UL 61010-2-010, UL 61010-2-051, CAN/CSA C22.2 No. 61010-1-12, CSA C22.2 No. 61010-2-010, CSA C22.2 No. 61010-2-051, IEC 61326-1, CISPR 11, FCC 47 CFR Part 15 (Class A)
Specifications

Specifications

9 to 15.8 kg
120 V 50/60 Hz
20°C Below Ambient to 80°C
6 L Flasks
20 to 35 lb.
±0.25°C at 37°C
Aluminum Platform
20.3 in.
51.5 cm
46 x 46 cm (18 x 18 in.)
32.2 in.
81.8 cm
25 in.
63.5 cm
266 lb.
±1 rpm
LED Digital Electric Display
20 to 80%, Non-condensing
USB, Ethernet
Universal/Dedicated
3 Year
25 to 400 rpm
120 V
1500 VA (12.5 A)
Incubator Shaker
68°F below ambient to 176°F
60 Hz
29.3 in.
74.5 cm
51.5 x 51.7 x 47.7 cm (20.3 x 20.4 x 18.8 in.)
63.5 x 74.5 x 81.8 cm / 25 x 29.3 x 32.8 in.
18.8 in.
47.7 cm
20.4 in.
51.7 cm
121 kg
Innova 42R
Plasmid DNA Production and Protein Expression, Biofuel Research, Grow Bacteria, Yeast, Fungus, Photosynthetic Organisms
Accessories
Universal/Dedicated
0.01 sec. to 95 hr. 59 min., Continuous
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.