Description
- Inserts to help retain tubes and prevent them from spinning in the rack
- Finger-tight knobs to quickly adjust angle, or release the rack from the base plate, for easy transport to the bench
- Stainless-steel
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Test Tube Rack |
| Material | Stainless Steel |
| Diameter (Metric) | 26 to 30 mm |
| For Use With (Equipment) | Innova 43/43R Incubator shaker |
| Includes | Two adjustable bars and non-skid rubber mat |
| Size | Small |