Description
- Inserts to help retain tubes and prevent them from spinning in the rack
- Finger-tight knobs to quickly adjust angle, or release the rack from the base plate, for easy transport to the bench
- Stainless-steel
Specifications
Specifications
| Product Type | Test Tube Rack |
| Material | Stainless Steel |
| Diameter (Metric) | 18 to 21 mm |
| For Use With (Equipment) | Innova 43/43R Incubator shaker |
| Includes | Two adjustable bars and non-skid rubber mat |
| Size | Medium |