Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Methylene Blue 'N' Stain, for Reticulocyte Count, Ricca Chemical
SDP

Catalog No. 52854 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

For reticulocyte count

Chemical Identifiers

CAS 6586-05-6, 6487-48-5, 7647-14-5, 6586-05-6
Molecular Formula C18H22Cl3N3SZn
Molecular Weight (g/mol) Mixture
InChI Key GGKWUSWRGRCLEV-UHFFFAOYSA-K
IUPAC Name zinc(2+) 3,7-bis(ethylamino)-2,8-dimethyl-5λ4-phenothiazin-5-ylium trichloride
SMILES [Cl-].[Cl-].[Cl-].[Zn++].CCNC1=CC2=[S+]C3=CC(NCC)=C(C)C=C3N=C2C=C1C

Specifications

CAS Min % 95.0, 1.34, 0.76, 0.43
CAS Max % 99.0, 1.39, 0.8, 0.45
Grade Laboratory
Packaging Natural Poly Bottle
Color Dark Blue
Quantity 120 mL
Chemical Name or Material New Methylene Blue ';N';
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.