Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Newcastle Disease Virus (NDV) Antibody (9F7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11059912
Description
Newcastle Disease Virus (NDV) Monoclonal specifically detects Newcastle Disease Virus (NDV) in Virus samples. It is validated for Western Blot, Haemagglutination, Sandwich ELISA.Specifications
Newcastle Disease Virus (NDV) | |
Monoclonal | |
Unconjugated | |
NDV | |
Newcastle disease virus (La-sota strain) | |
0.2 mg | |
Primary | |
Virus | |
Purified |
ELISA | |
9F7 | |
Sandwich ELISA 1:100-1:2000 | |
Mouse | |
Protein G purified | |
RUO | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunised with Newcastle disease virus (La-sota strain). | |
Store at 4C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction