Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ Synthetic Blend Mops

Catalog No. 19050660 Shop All Newell Rubbermaid Inc Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-050-660 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-050-660 Supplier Rubbermaid™ Supplier No. FGF13600BL00

Specifications

Product Type Mop
Color Blue
Length (English) 7.5 in.
Width (English) 6.5 in.
For Use With (Application) For General Purpose
Material Synthetic Blend
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.