Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ Universal Headband Cotton Mops

Catalog No. 19120407 Shop All Newell Rubbermaid Inc Products
Change view
Click to view available options
Capacity:
16 oz.
24 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
19-120-407 16 oz.
19-120-408 24 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-120-407 Supplier Rubbermaid™ Supplier No. FGE13600WH00

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Product Type Headband Mop
Color White
Capacity 16 oz.
Quantity 12/Cs.
For Use With (Application) For General Cleaning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.