Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Learning I Love Math Hands-On Program
Shop All NewPath Learning Products

Click to view available options
Grade:
1
2
3
4
5 to 12
Description
Complete coverage of 15 concepts: Whole Numbers, Patterns and Algebra, Mass, Addition and Subtraction, Data, Time, Multiplication and Division, Length, Three-Dimensional Space, Chance, Volume and Capacity, Position
- Developed by an experienced educator and classroom-tested for more than a decade
- Wealth of challenging, mathematical exercises focused on leveled, standards-based topics
- Fun-filled means of motivating students to think more deeply, investigate, explain, and understand problem-solving strategies
- Concepts divided into two sections: one per semester
- Each section is two to three pages in length and followed by an assessment for immediate and continuous feedback
Specifications
Specifications
| Grade Level | Elementary |
| Grade | 4 |
| Format | E-books/Workbooks |
| Product Type | I Love Hands-on Math Reproducible Workbook |
| For Use With (Application) | Fun filled means of motivating students to think more deeply, investigate, explain and understand problem-solving strategies |
| Subject | Mathematics |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction