Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

What are Ecosystems? Learning Guide
SCI_ED

Catalog No. S13893 Shop All NewPath Learning Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S13893 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13893 Supplier NewPath Learning Supplier No. 143260
Add to Cart
Add to Cart

Use engaging graphics and inquiry-based activities to enhance student comprehension on standards-based concepts on Ecosystems

Full-color, 36-page guide features self-directed readings, easy-to-follow illustrated explanations; guiding questions; inquiry-based activities; a lab investigation; key vocabulary review; and assessment review questions along with a post-test

Topics Covered:

  • Environment
  • Living and Nonliving Things
  • What is an Ecosystem?
  • An Organism's Habitat and Niche
  • Population and Community
  • Parts of an Ecosystem
  • How Does an Ecosystem Work?
  • Kinds of Ecosystems
  • Energy in an Ecosystem
  • Producers, Consumers and Decomposers
  • How Do Ecosystems Change?
  • Environmental Changes and Effects on Organism

Specifications

Product Type Learning Guide
Format 36-page workbook
For Use With (Application) Enhance student comprehension on standards-based concepts on ecosystems
Certifications/Compliance Meets Academic Standard for California, Florida, National, New York, Texas and Next Generation Science Standards
Quantity 10/Pk.
Grade Level Elementary
Grade 3 to 5
Subject Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.