Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nexilin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP153226

Catalog No. NBP153226


Only null left
Add to Cart

Description

Description

Nexilin Polyclonal specifically detects Nexilin in Human samples. It is validated for Western Blot.
Specifications

Specifications

Nexilin
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
CMD1CC, F-actin-binding protein, MGC104234, MGC138865, MGC138866, Nelin, NELINnexilin, nexilin (F actin binding protein)
Rabbit
81 kDa
100 μL
Primary
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
Q0ZGT2
NEXN
Synthetic peptides corresponding to NEXN(nexilin (F actin binding protein)) The peptide sequence was selected from the middle region of NEXN. Peptide sequence EELERQRQENRKKQAEEEARKRLEEEKRAFEEARRQMVNEDEENQDTAKI.
Affinity purified
RUO
91624
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only