Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEXN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110024 Supplier Invitrogen™ Supplier No. PA5110024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NEXN Polyclonal Antibody for ELISA, ICC/IF, Western Blot

Involved in regulating cell migration through association with the actin cytoskeleton. Has an essential role in the maintenance of Z line and sarcomere integrity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEXN
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NEXN
Gene Accession No. Q0ZGT2, Q7TPW1, Q9Z2J4
Gene Alias 1110046H09Rik; AA553326; CMH20; F-actin binding protein; F-actin-binding protein; Nelin; Nexilin; nexilin (F actin binding protein); nexilin alpha; nexilin beta; nexilin F-actin binding protein; nexilin gamma; NEXN; nexn {ECO:0000312; nF actin binding protein; RGD:708354}
Gene Symbols NEXN
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246172, 68810, 91624
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.