Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEXN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552803 Supplier Invitrogen™ Supplier No. PA552803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEERARRRAI DLEIKEREAE NFHEEDDVDV RPARKSEAPF THKVNMKARF EQMAKAREEE EQRRIEEQKL LRMQFEQREI DAALQKKREE EEEEEGSIMN GSTAEDEEQT RSGA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.

Involved in regulating cell migration through association with the actin cytoskeleton. Has an essential role in the maintenance of Z line and sarcomere integrity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEXN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEXN
Gene Accession No. Q0ZGT2
Gene Alias 1110046H09Rik; AA553326; CMH20; F-actin binding protein; F-actin-binding protein; Nelin; Nexilin; nexilin (F actin binding protein); nexilin alpha; nexilin beta; nexilin F-actin binding protein; nexilin gamma; NEXN; nexn {ECO:0000312; nF actin binding protein; RGD:708354}
Gene Symbols NEXN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEXN. Recombinant protein control fragment (Product #RP-90647).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91624
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.