Learn More
Nextteq™ Gastec™ Detector Tubes: Acetone
SDP
Catalog No. 19026196 Shop All Nextteq Products
Change view
Click to view available options
Measurement Range:
50 to 12000
Catalog No. Measurement Range
19-026-196 50 to 12000
1 options
This item is not returnable. View return policy
Catalog No. 19-026-196 Supplier Nextteq™ Supplier No. 151L
This item is not returnable. View return policy
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
Components Measured Acetone
Measurement Range 50 to 12000
No. of Strokes 1 to 2
Quantity 10
Response Time 2 to 4 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C