Learn More
Nextteq™ Gastec™ Detector Tubes: Formaldehyde
SDP
Catalog No. 19026356 Shop All Nextteq Products
Change view
Click to view available options
Components Measured:
Formaldehyde
Formaldehyde Low Range
Measurement Range:
0.05 to 1
0.1 to 40
2 to 100
Response Time:
1.5 to 7.5 min.
30 sec. to 1.5 min.
7.5 min.
No. of Strokes:
0.5 to 2
1 to 5
5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range Response Time No. of Strokes
19-026-356 Formaldehyde 2 to 100 30 sec. to 1.5 min. 0.5 to 2
19-026-359 Formaldehyde Low Range 0.05 to 1 7.5 min. 5
19-026-358 Formaldehyde 0.1 to 40 1.5 to 7.5 min. 1 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-026-356 Supplier Nextteq™ Supplier No. 91
Add to Cart
Add to Cart
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
Components Measured Formaldehyde
Type Gas Detection Tube
Measurement Range 2 to 100
No. of Strokes 0.5 to 2
Quantity 10
Response Time 30 sec. to 1.5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C