Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nextteq™ Gastec™ Detector Tubes: Methyl Mercaptan
SDP

Catalog No. 19026327 Shop All Nextteq Products
Change view
Click to view available options
Measurement Range:
0.25 to 140ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range
19-026-327 0.25 to 140ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-026-327 Supplier Nextteq™ Supplier No. 71
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.

Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.

Specifications

Components Measured Methyl Mercaptan
Type Gas Detection Tube
Measurement Range 0.25 to 140ppm
No. of Strokes 0.5 to 10
Quantity 10
Response Time 45 sec. to 15 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.