Learn More
Nextteq™ Gastec™ Dosimeter Tubes
SDP
Catalog No. 19026275 Shop All Nextteq Products
Change view
Click to view available options
Components Measured:
Acetaldehyde, Formaldehyde, Furfural, Methyl Ethyl Ketone
Ammonia, Dimethylamine, N,N-Diethylmethylamine, Hydrazine, Triethylamine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19-026-275 Ammonia, Dimethylamine, N,N-Diethylmethylamine, Hydrazine, Triethylamine
19-026-357 Acetaldehyde, Formaldehyde, Furfural, Methyl Ethyl Ketone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-026-275 Supplier Nextteq™ Supplier No. 3D
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Components Measured Ammonia, Dimethylamine, N,N-Diethylmethylamine, Hydrazine, Triethylamine
Measurement Range Ammonia: 2.5-1000ppm, Dimethylamine: 1.9-750ppm, N,N-Diethylmethylamine: 4-1600ppm, Hydrazine: 1.5-650ppm, Triethylamine: 5.3-2100ppm
Quantity 10
Response Time Ammonia: 0.5-10 hr., Dimethylamine: 0.5-10 hr., N,N-Diethylmethylamine: 0.5-10 hr., Hydrazine: 0.5-10 hr., Triethylamine: 0.5-10 hr.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C