Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NF 110
GREENER_CHOICESDP

Catalog No. 254810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-481-0 10 mg
25-485-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-481-0 Supplier Tocris Bioscience™ Supplier No. 2548/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent P2X3 antagonist

High affinity P2X3 receptor antagonist (Ki values are 36, 82 and 4144 nM for P2X3, P2X1 and P2X2 recombinant receptors respectively). Shows no activity at P2Y1, P2Y2 and P2Y11 receptors (IC50 > 10 μM). Potently inhibits α,β-meATP-evoked desensitizing currents in rat DRG neurons (IC50 = 527 nM). Shows antitumor activity against several tumor types. Also inhibits DNA-binding activity of HMGA2 (IC50 = 0.87 μM).

Chemical Identifiers

CAS 111150-22-2
Molecular Formula C41H28N6Na4O17S4

Specifications

Quantity 10 mg
Formula Weight 1096.9
Purity >98%
Molecular Weight (g/mol) 1096.899
Chemical Name or Material NF 110
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.