Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NF 157
GREENER_CHOICESDP

Catalog No. 245010 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-501-0 Supplier R&D Systems™ Supplier No. 2450/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective P2Y11 and P2X1 antagonist

NF 157 is a purinergic receptor antagonist that potently inhibits P2Y11 receptor activity (IC50 = 463 nM). Displays selectivity for P2Y11 and P2X1 receptors over P2Y1, P2Y2, P2X2, P2X3, P2X4 and P2X7 receptors. Inhibits NAD+-induced activation of human granulocytes.

This product is supplied with a high degree of hydration and some residual NaCl, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content and the maximum solubility threshold to use in dilution calculations.

Chemical Identifiers

CAS 104869-26-3
Molecular Formula C49H28F2N6Na6O23S6

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 1437.08
Purity 90%
Molecular Weight (g/mol) 1437.077
Chemical Name or Material NF 157
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.