Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-1A Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501994026
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4026 100 μL
50-199-4027 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4026 Supplier Active Motif Supplier No. 39397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.

Specifications

Antigen NF-1A
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene NFIX
Gene Accession No. Q14938
Gene Alias CCAAT-binding transcription factor; CCAAT-box-binding transcription factor; CTF; KIAA1439; MRSHSS; NF1A; NF1-A; NF1-B; NF1-C; NF1-X; NF1-X1 protein; NFI; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; NFIB; NFI-B; NFIC; NFI-C; Nfix; NFI-X; Nuclear factor 1 X-type; Nuclear factor 1/A; Nuclear factor 1/B; Nuclear factor 1/C; nuclear factor 1/X; nuclear factor I X; Nuclear factor I/A; Nuclear factor I/B; Nuclear factor I/C; Nuclear factor I/X; nuclear factor I/X (CCAAT-binding transcription factor); SOTOS2; TGGCA-binding protein
Gene Symbols NFIX
Host Species Rabbit
Immunogen This NF-1A antibody was raised against a synthetic peptide corresponding to amino acid residue(s) 478-492 of human NF-1A.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4784
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.