Learn More
R&D Systems™ NFκB p50 Transcription Factor Assay
GREENER_CHOICESDP
Catalog No. 705125 Shop All Novus Products
Change view
Click to view available options
Quantity:
25 mg
Catalog No. Quantity
70-512-5 25 mg
1 options
This item is not returnable. View return policy
Catalog No. 70-512-5 Supplier R&D Systems™ Supplier No. 7051/25
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
  • Transcription Factor Assay Plate
  • TFA Buffer, 5x
  • Blocking Reagent
  • Blocking Reagent
  • Primary Antibody
  • Gt X Rb, HRP Secondary Antibody
  • NFkB Capture Probe
  • NFkB Specific Competitor Oligonucleotide
  • TFA Negative Control Probe
  • HeLa Whole Cell Extract, TNFalpha Treated
  • Substrate (TMB/E)
  • Stop Solution
A Tocris product, part of R&D Systems. The transcription factor NFkB is involved in the expression and regulation of a number of important cellular and physiological processes such as growth, development, apoptosis, immune and inflammatory response
Tasquinimod is a high affinity negative allosteric modulator of HDAC4 (Kd = 10 - 30 nM). Binds the regulatory Zn2+ binding domain of HDAC4. Suppresses hypoxia-induced decrease in histone acetylation in human prostate cancer cells in vitro. Also binds S100A9. Antiangiogenic. Inhibits endothelial sprouting in vitro and growth of prostate tumor xenografts in nude mice. Orally bioavailable.
Specifications
Quantity 25 mg
Product Line Tocris