Learn More
R&D Systems™ Tasquinimod
GREENER_CHOICESDP
Catalog No. 70515 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Plate
Catalog No. Quantity
70-515 1 Plate
1 options
This item is not returnable. View return policy
Catalog No. 70-515 Supplier R&D Systems™ Supplier No. 7051/5
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Tasquinimod is a high affinity negative allosteric modulator of HDAC4 (Kd = 10 to 30 nM).
Tasquinimod is a high affinity negative allosteric modulator of HDAC4 (Kd = 10 - 30 nM). Binds the regulatory Zn2+ binding domain of HDAC4. Suppresses hypoxia-induced decrease in histone acetylation in human prostate cancer cells in vitro. Also binds S100A9. Antiangiogenic. Inhibits endothelial sprouting in vitro and growth of prostate tumor xenografts in nude mice. Orally bioavailable.
Specifications
Quantity 1 Plate
Product Line Tocris