Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-H Antibody (AH1), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $512.00
Specifications
Antigen | NF-H |
---|---|
Clone | AH1 |
Concentration | 1 mg/ml |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NF-H Monoclonal specifically detects NF-H in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Specifications
NF-H | |
1 mg/ml | |
Unconjugated | |
Mouse | |
Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
NEFH | |
IgG1 | |
Affinity purified | |
Clone AH1 recognizes phosphorylated NF-H KSP sequences. Does not recognize non-phosphorylated KSP sequences. |
AH1 | |
Monoclonal | |
Ascites | |
RUO | |
Pig, Rat, Bovine, Equine, Human, Mouse | |
4744 | |
Native axonal phosphorylated Neurofilament Heavy protein purified from bovine spinal cord | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
200 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title