| Antigen |
NF-H |
| Applications |
Flow Cytometry, Immunohistochemistry (Paraffin), CyTOF |
| Classification |
Monoclonal |
| Clone |
NE14 |
| Concentration |
1.0 mg/mL |
| Conjugate |
Unconjugated |
| Dilution |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.25 - 0.5 ug/ml, CyTOF-ready |
| Formulation |
PBS with No Preservative |
| Gene Alias |
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H |
| Gene Symbols |
NEFH |
| Host Species |
Mouse |
| Immunogen |
Crude neurofilament preparation from porcine spinal cord |
| Molecular Weight of Antigen |
200 kDa |
| Purification Method |
Protein A or G purified |
| Quantity |
0.1 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
4744 |
| Test Specificity |
This MAb reacts with a 200kDa protein, identified as heavy sub-unit of neurofilaments (NF-H). It reacts specifically with the phosphorylated KSP/KEP segment at the C-terminus of the heavy subunit (NF-H) of neurofilaments. After dephosphorylation of neurofilaments with alkaline phosphatase, this Ab no longer binds. Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68kDa (NF-L), 160kDa (NF-M) and 200kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament. |
| Target Species |
Human, Mouse, Rat, Pig, Chicken, Feline, Gerbil, Guinea Pig, Primate, Rabbit |
| Content And Storage |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Form |
Purified |
| Isotype |
IgG1 |
|
Show More
Show Less
|
|