Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-H Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | NF-H |
---|---|
Dilution | Western Blot 1:10000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:2000 |
Applications | Immunohistochemistry (Free Floating), Immunocytochemistry, Immunofluorescence, ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NF-H Polyclonal specifically detects NF-H in Rat, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Specifications
NF-H | |
Immunohistochemistry (Free Floating), Immunocytochemistry, Immunofluorescence, ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
Whole antisera with No Preservative | |
4744 | |
Purified rat Neurofilament Heavy construct containing most of the tandem KSP repeats [UniProt# P16884] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:10000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
Rat, Chicken | |
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
NEFH | |
IgG | |
Unpurified | |
200 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title