Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-H Antibody (RT-97 + NR-4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247664PCP
Description
NF-H Monoclonal specifically detects NF-H in Human, Mouse, Rat, Porcine, Chicken samples. It is validated for Flow Cytometry.Specifications
NF-H | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
NEFH | |
Triton-X 100 insoluble proteins of rat brain (RT-97); Neurofilaments from porcine spinal cord (NR-4) (Uniprot: P12036) | |
0.1 mL | |
Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
This monoclonal antibody reacts with a 200kDa and 68kDa protein, identified as heavy and light sub-units of neurofilaments (NF-H NF-L). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68kDa (NF-L), 160kDa (NF-M) and 200kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament. | |
Store at 4°C in the dark. |
Flow Cytometry | |
RT-97 + NR-4 | |
Flow Cytometry | |
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Chicken | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction