Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-H Antibody (SPM203) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2478330.2MG
Description
NF-H Monoclonal antibody specifically detects Antigen in Chicken, Human, Mouse, Porcine, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
NF-H | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry, Immunohistochemistry-Paraffin : 0.25 - 0.5 ug/ml, CyTOF-ready | |
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
Mouse | |
200 kDa | |
0.2 mg | |
Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
4744 | |
Human, Mouse, Rat, Pig, Chicken | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
SPM203 | |
Unconjugated | |
PBS with No Preservative | |
NEFH | |
Triton-X 100 insoluble protein fraction of rat brain | |
Protein A or G purified | |
RUO | |
Primary | |
This MAb reacts with a 200kDa protein, identified as heavy sub-unit of neurofilaments (NF-H). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68kDa (NF-L), 160kDa (NF-M) and 200kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction