Learn More
Invitrogen™ NF-H Monoclonal Antibody (NF-05)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA110441
Description
This antibody recognizes a nonphosphorylated epitope of neurofilament heavy protein (NF-H), a 210 kDa intracellular structural protein of Intermediate Filament Proteins family. NF-H is mainly expressed in the central and peripheral nervous system and reproductive system and is biochemically very stable. It will not cross-react with mouse.
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
Specifications
NF-H | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P12036, P12037, P16884 | |
Nefh | |
Pig brain neurofilament protein-enriched fraction after depolymerization of microtubules. | |
100 μg | |
Primary | |
Human, Rat, Pig | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Frozen), Western Blot | |
NF-05 | |
Unconjugated | |
Nefh | |
200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg | |
Mouse | |
Sequential Chromatography | |
RUO | |
100156492, 24587, 4744 | |
4°C, do not freeze | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.