Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NF-H Monoclonal Antibody (RNF 402)
GREENER_CHOICE

Catalog No. OMA106116
missing translation for 'changeView'
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OMA106116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. OMA106116 Supplier Invitrogen™ missing translation for 'supplierNo' OMA106116
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OMA1-06116 detects both the phosphorylated and non-phosphorylated forms of the 200 kDa neurofilament from rat, guinea pig, rabbit, dog, hamster, chicken, mouse, cow, monkey, sheep, xenopus and human tissues. OMA1-06116 has been successfully used in immunohistochemical (frozen & paraffin) and Western blot procedures. By Western blot, this antibody detects a single ~200 kDa protein representing both forms of the 200 kDa neurofilament from rat brain homogenate. OMA1-06116 antigen is cytoskeletal preparation of calf brain tissue.

Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-H
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone RNF 402
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Nefh
Gene Accession No. P12036, P16884, P19246
Gene Alias 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg
Gene Symbols Nefh, nefh.L
Host Species Mouse
Immunogen Neurofilament preparation of calf brain tissue.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100101753, 100357177, 100716406, 100753127, 101107472, 101838568, 108715402, 24587, 380684, 417020, 442940, 4744, 528842
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Chicken, Guinea Pig, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.