Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NF-H Monoclonal Antibody (RNF 405)
GREENER_CHOICE

Catalog No. OMA106117
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OMA106117 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OMA106117 Supplier Invitrogen™ Supplier No. OMA106117
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OMA1-06117 reacts exclusively with the phosphorylated form of the 200 kDa neurofilament from human, mouse, rat, canine, rabbit, monkey, bovine, ovine, hamster, chicken, guinea pig, and Xenopus laevis tissues. OMA1-06117 has been successfully used in Western blot and IHC (P) procedures. By Western blot, this antibody detects a single ~200 kDa protein representing phosphorylated neurofilament from rat brain homogenate. OMA1-06117 antigen is cytoskeletal preparation of calf brain tissue.

Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-H
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RNF 405
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Nefh
Gene Accession No. P12036, P16884, P19246
Gene Alias 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg
Gene Symbols Nefh, nefh.L
Host Species Mouse
Immunogen Neurofilament preparation of calf brain tissue.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100101753, 100357177, 100716406, 100753127, 101107472, 101838568, 108715402, 24587, 380684, 417020, 442940, 4744, 528842
Target Species Bovine, Canine, Chicken, Guinea Pig, Hamster, Human, Mouse, Monkey, Sheep, Rabbit, Rat, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.