Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-H Antibody (RNF405), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197680AF532
Description
NF-H Monoclonal specifically detects NF-H in Human, Mouse, Rat, Bovine, Canine, Chicken, Guinea Pig, Hamster, Primate, Monkey, Rabbit, Sheep, Xenopus, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
NF-H | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
NEFH | |
Clone RNF405 is a mouse monoclonal IgG2a antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a mouse immunized with a neurofilament preparation of calf brain tissue. | |
Primary | |
RNF405 reacts exclusively with the phosphorylated isoform of the 200 kD neurofilament protein. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
RNF405 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting | |
200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
Mouse | |
0.1 mL | |
4744 | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Guinea Pig, Hamster, Primate, Monkey, Rabbit, Sheep, Xenopus species, Zebrafish | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction