Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-L Antibody (7D1), Novus Biologicals™
SDP

Catalog No. NBP250612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250612 0.1 mL
NB019697 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP250612 Supplier Novus Biologicals Supplier No. NBP250612
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NF-L Monoclonal specifically detects NF-L in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen NF-L
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:10000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Alias 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa
Gene Symbols NEFL
Host Species Mouse
Immunogen A preparation of pig Neurofilament Light and Neurofilament Heavy
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4747
Target Species Human, Mouse, Rat, Pig, Bovine, Mammalia
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.