Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-L Antibody (DA2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$243.50 - $353.50
Specifications
Antigen | NF-L |
---|---|
Clone | DA2 |
Dilution | Western Blot 1:5000 - 1:10000, Simple Western 1:50, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25565998) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300132SS
![]() |
Novus Biologicals
NB300132SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB300132
![]() |
Novus Biologicals
NB300132 |
0.05 mL |
Each for $353.50
|
|
|||||
Description
NF-L Monoclonal specifically detects NF-L in Human, Mouse, Rat, Avian, Bovine, Mammal samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NF-L | |
Western Blot 1:5000 - 1:10000, Simple Western 1:50, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25565998) | |
Monoclonal | |
Ascites | |
RUO | |
Human, Mouse, Rat, Avian, Bovine, Mammalia | |
68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
NEFL | |
IgG1 κ | |
Unpurified | |
Specifically recognizes the light neurofilament subunit or NF-L (∼68 kDa). |
DA2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
Ascites with 0.1% Sodium Azide | |
4747 | |
Native Neurofilament Light protein purified from pig spinal cord [UniProt# P02547] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title