Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-L Antibody (NFL/736), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2448930.2MG
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Immunofluorescence
NF-L Monoclonal antibody specifically detects NF-L in Human, Rat, Porcine, Chicken, Primate samples. It is validated for Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| NF-L | |
| Monoclonal | |
| 0.2mg/mL | |
| Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
| P07196 | |
| NEFL | |
| Recombinant human NEFL protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This MAb reacts with a 68kDa protein, identified as light sub-unit of neurofilaments (NF-L). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68kDa (NF-L), 160kDa (NF-M) and 200kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament. | |
| Store at 4C. | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunofluorescence | |
| NFL/736 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
| Mouse | |
| 68 kDa | |
| 0.2 mg | |
| Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
| 4747 | |
| Human, Rat, Pig, Chicken, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction