Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-L Antibody, Novus Biologicals™
SDP

Chicken Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen NF-L
Dilution Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000
Applications Immunohistochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018362
SDP
View Documents
Novus Biologicals
NBP1052190.025ML
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP105219
SDP
View Documents
Novus Biologicals
NBP105219
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

NF-L Polyclonal specifically detects NF-L in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Specifications

Specifications

NF-L
Immunohistochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
Chicken
Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience
P02548
4747
Recombinant human NF-L protein
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000
Polyclonal
Precipitation
RUO
Mouse, Human, Bovine, Rat, Pig
68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa
NEFL
IgY
Ammonium sulfate precipitation
68 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.