Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-M Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
Supplier: Novus Biologicals NB300133
Description
NF-M Polyclonal specifically detects NF-M in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
NF-M | |
Polyclonal | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:1000-1:2500, Immunocytochemistry/Immunofluorescence 1:1000-1:2500, Immunohistochemistry-Paraffin 1:1000-1:2500, Immunohistochemistry-Frozen 1:1000-1:2500, Knockout Validated | |
P12839 | |
NEFM | |
Recombinant fusion protein containing the extreme C-terminal segment of rat NF-M, amino acids 549-845 | |
Unpurified | |
RUO | |
Primary | |
Specifically recognizes the evolutionarily conserved extreme C-terminal region of Neurofilament Medium (∼145-170kDa). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Supplied as serum with 5mM Sodium Azide | |
NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein | |
Rabbit | |
160 kDa | |
0.1 mL | |
Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
4741 | |
Human, Mouse, Rat, Pig, Bovine, Chicken, Equine | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction