Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-M Rabbit anti-Human, Clone: 5E0D2, Novus Biologicals™
SDP

Catalog No. NB166405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166405 100 μg
NB166406 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166405 Supplier Novus Biologicals Supplier No. NBP315655100UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NF-M Monoclonal antibody specifically detects NF-M in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen NF-M
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 5E0D2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 600-700 of human NF-M (P07197).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4741
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.