Learn More
NF Mouse anti-Human, Clone: OTI3F7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF190077

Description
Synthetic peptide sequence is EYQDLLNVKMALDIEIAAYRKLLEGEE Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the medium neurofilament protein. This protein is commonly used as a biomarker of neuronal damage. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Diseases associated with NEFM include Pineal Parenchymal Tumor Of Intermediate Differentiation and Wallerian Degeneration. Among its related pathways are Amyotrophic lateral sclerosis (ALS) and Neural Stem Cell Differentiation Pathways and Lineage-specific Markers. Gene Ontology (GO) annotations related to this gene include structural molecule activity and structural constituent of cytoskeleton.Specifications
NF | |
Monoclonal | |
Unconjugated | |
NEFL | |
RNA5-8N2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4741, 4744, 4747 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3F7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P07196, P07197, P12036 | |
NEFH, NEFL, NEFM | |
Synthetic peptide corresponding to the conserved region of human neurofilament heavy polypeptide (NEFH), medium polypeptide (NEFM) and light polypeptide (NF68, NEFL) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.