Learn More
NF-YC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | NF-YC |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2336
![]() |
Bethyl Laboratories, Inc
IHC00577T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2335
![]() |
Bethyl Laboratories, Inc
IHC00577 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Guinea pig_10141, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Northern whi
The CCAAT-binding factor NF-Y is a heteromeric transcription factor that specifically binds to CCAAT sequences in many eukaryotic genes. NF-Y is made up of three subunits, NF-YA, NF-YB and NF-YC. All three components are necessary for DNA binding. In each NF-Y subunit, the segment needed for formation of the NF-Y-DNA complex is conserved from yeast to human. These conserved segments are homologous to the histone-fold motif of eukaryotic histones. The DNA binding domains of the NF-YB and NF-YC subunits have been suggested to interact through a protein-protein histone-fold “handshake” motif in a manner analogous to the histone proteins, H2B and H2A, respectively.Specifications
NF-YC | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Nuclear transcription factor Y subunit gamma, CAAT box DNA-binding protein subunit C, Nuclear transcription factor Y subunit C, NF-YC, Transactivator HSM-1/2, NFYC | |
Between 408 and 458 | |
Primary | |
4° C | |
NFYC |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P70353, Q13952 | |
18046, 4802 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.