Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF2/Merlin Antibody (534428) [HRP], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5616H
Description
NF2/Merlin Monoclonal antibody specifically detects NF2/Merlin in Human samples. It is validated for Western BlotSpecifications
NF2/Merlin | |
Monoclonal | |
HRP | |
ACN, BANF, merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin | |
E. coli-derived recombinant human NF2/Merlin, Met363-Lys578, Accession # P35240 | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Phospho Specific, Tumor Suppressors | |
4771 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot | |
534428 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction