Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF2/Merlin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7202543 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092634 20 μL
NB092633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092634 Supplier Novus Biologicals Supplier No. NBP30374020UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NF2/Merlin Polyclonal specifically detects NF2/Merlin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen NF2/Merlin
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ACN, BANF, merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 477-576 of human NF2/Merlin (NP_000259.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4771
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.