Learn More
NF90/NF110 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303120AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
NF90 (nuclear factor of activated T-cells 90 kDa), also known as NFAR, DRBF, DRBP76 (double-stranded RNA-binding protein 76), MPP4, MPHOSPH4 (M-phase phosphoprotein 4), ILF3 (interleukin enhancer binding factor 3) or TCP80 (translational control protein 80), is a ubiquitously expressed nuclear protein that exists in a heterodimer with NF45. NF90 contains two DRBM (double-stranded RNA(dsRNA-)-binding) domains and one DZF domain and, in association with NF45 primarily participates in the regulation of IL-2 expression by binding to the antigen receptor response element (ARRE) target sequence of the IL-2 enhancer. In neuronal cells, the NF45/NF90 heterodimer can repress human rhinovirus type 2 replication by binding to a 5′ untranslated region of the viral RNA that encodes the internal ribosome entry site (IRES). NF45 and NF90 belong to the double-stranded RNA-binding protein family and both are substrates for the dsRNA-activated protein kinase, PKR. Due to alternative splicing events, six isoforms exist for NF90, namely NFAR-2 (or ILF3-E), NFAR-1 (or DRBP76), isoform 3, DRBP76 alpha (or ILF3-A), DRBP76 delta (also known as DRBP76 gamma or ILF3-C) and isoform 6.Specifications
NF90/NF110 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q12906 | |
ILF3 | |
Between 50 and 100 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
ILF3 | |
Interleukin enhancer-binding factor 3, Double-stranded RNA-binding protein 76, DRBP76, M-phase phosphoprotein 4, MPP4, Nuclear factor associated with dsRNA, NFAR, Nuclear factor of activated T-cells 90 kDa, NF-AT-90, Translational control protein 80, TCP80, DRBF, MPHOSPH4, NF90 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3609 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.