Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103469 Supplier Invitrogen™ Supplier No. PA5103469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NFAM1.

The protein encoded by this gene is a type I membrane receptor that activates cytokine gene promoters such as the IL-13 and TNF-alpha promoters. The encoded protein contains an immunoreceptor tyrosine-based activation motif (ITAM) and is thought to regulate the signaling and development of B-cells. It is also known CNAIP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFAM1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NFAM1
Gene Accession No. Q8NET5, Q8R4V1
Gene Alias 4921501M20Rik; bK126B4.4; Calcineurin/NFAT-activating ITAM-containing protein; Cnaip; CTA-126B4.4; Nfam1; Nfat activating molecule with ITAM motif 1; NFAT activating protein with ITAM motif 1; NFAT activation molecule 1; NFAT-activating protein with ITAM motif 1
Gene Symbols NFAM1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NFAM1(Accession Q8NET5), corresponding to amino acid residues L183-C233.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150372, 74039
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.